Lab-Aids® DNA and its Replication Model Kit
# de catalogue 470123-044
Fournisseur: Lab-Aids
Spécifications
- Description:DNA And Its Replication Model Kit
- No. of Lab Groups:12
- Cat. no.:470123-044
- Supplier No.:71
Spécifications
A propos de cet article
A basic introduction to DNA.
- Build a segment of DNA
- Unzip and replicate
- Reusable
- Time required: one 50 minute class period
Packagaged as 12 individually segments for classroom use of 12 students or teams. Students will build models of up to 8 BP in length and turn into a helix. Individual pieces replicate basic DNA structure. The segments can be attached together to make a giant double helix over 2.7 M long, and can be 'unzipped' to permit students to replicate their segment. The reusable, color-coded, plastic components (672 pieces) are easily assembled or disassembled.
Delivery: See datasheet for contents information.
Customers Who Bought This Also Bought