ContactUsLinkComponent
Ward's® DNA Amplification by PCR Lab Activity
Catalogue # 470024-478
Fournisseur:  Avantor
undefined
Ward's® DNA Amplification by PCR Lab Activity
Catalogue # 470024-478
Fournisseur:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    DNA Amplification by PCR Kit
  • No. of Lab Groups:
    8
  • Cat. no.:
    470024-478
Spécifications
À propos de cet article
Introduce Students to PCR Technique with a Basic Procedure
  • Reliable PCR reaction
  • Materials for 24 PCR reactions
  • Time Required: ~60 min. plus 2 hrs. for PCR
In this investigation, students amplify various segments of the bacteriophage lambda genome, then electrophorese and analyze the PCR samples. Students use the sizes of the PCR products generated by the three different forward primers and one reverse primer, provided, to map the locations of the primers relative to one another in the bacteriophage lambda genome. You will get enough materials to run up to 24 PCR reactions, as well as a teacher’s guide and student copymaster.
Ordering information: Kit includes 5 mL of sterile Mineral Oil; 5 mL of sterile Water; 500mL of 5X TBE Running Buffer; 0.5 mL vial of Loading Dye; 200ml of 2% Agarose Gel; 24 X 0.5 mLPlastic Reaction Tubes; Floating Rack; 60mL bottle of Ward’s QUIKView DNA Stain Concentrate; biohazard bag; coupon for perishable material. Materials needed but not provided are electrophoresis Chamber; power supply; water bath or thermal cycler; micropipettes; marking pens;; timer; UV Transilluminator; UV Goggles; and ice. Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.