ContactUsLinkComponent
Dissectible Coils and Cores
  470120-754
TRANSFORMER DEMOUNTABLE
Dissectible Coils and Cores
  470120-754
  • Description:
    Dissectible Coils and Cores
  • Cat. no.:
    470120-754
  • Supplier No.:
    PH0850CSET
This set features five coils.
  • Coils Illustrate Electromagnetism and Induction
  • Useful in Basic Transformer Experiments
  • Included Coils Have 200, 400, 800, 1600, and 3200 Turns
  • Five Coils in Set
The coils illustrate electromagnetism and induction. They are also useful in basic transformer experiments.
The included coils have 200, 400, 800, 1600, and 3200 turns.
Customers Who Bought This Also Bought