Direct Reading Anemometer, AMERICAN SCIENTIFIC LLC SE
# de catalogue 470101-782
Fournisseur: American Scientific
Small Business Enterprise
Spécifications
- Description:Anemometer
- Cat. no.:470101-782
- Supplier No.:6577-00
Spécifications
A propos de cet article
As the wind blows, the arms on this unique anemometer move and lift the reading bar, allowing your students to get a direct reading of wind speed at just a glance. The colorful plastic instrument can be hand-held or permanently mounted. It measures the Beaufort scale from 0–11. Size: 30 x 18cm.
Customers Who Bought This Also Bought