Diffraction Grating Glasses
# de catalogue 470015-970
Fournisseur: RAINBOW SYMPHONY INC SE
Spécifications
- Description:Diffraction Grating Glasses
- Cat. no.:470015-970
- Supplier No.:01101
- No. of Parts:6
Spécifications
A propos de cet article
These glasses make the study of light and color fun and easy.
- Double-Axis Glasses
- Lenses Separate Light from Other Sources
- Economical
13,500 lines per inch double-axis holographic grating lenses separate light from any source into its spectral components for study and analysis.
Glasses are priced inexpensively enough to outfit your entire class.
Packaging: One package contains six glasses.
Customers Who Bought This Also Bought