Diatom Strew Set
# de catalogue 470026-846
Fournisseur: Avantor
Spécifications
- Description:Diatom Strew Set Fossil Specimen
- Cat. no.:470026-846
- Supplier No.:WARD470026-846
Spécifications
A propos de cet article
It has set of four transparent diatom strew slides: fossil freshwater, fossil marine, recent freshwater, and recent marine.
Customers Who Bought This Also Bought