ContactUsLinkComponent
Determination of Protein Molecular Weight Kit (EDVO-Kit 153)
Determination of Protein Molecular Weight Kit (EDVO-Kit 153)
# de catalogue 470120-898
Fournisseur:  Edvotek
DETERMINATION PROTEIN MW
Determination of Protein Molecular Weight Kit (EDVO-Kit 153)
# de catalogue 470120-898
Fournisseur:  Edvotek
Spécifications
  • Description:
    Determination of Protein Molecular Weight Kit
  • Cat. no.:
    470120-898
  • Supplier No.:
    153-B
Spécifications
A propos de cet article
Using pre-stained LyphoProteins™, analysis with denaturing SDS vertical polyacrylamide gel electrophoresis determines subunit molecular weights.
  • Material for Six Lab Groups Sharing Three Gels
  • Lab Uses Pre-Stained LyphoProteins
  • Vertical Electrophoresis Apparatus Required
Pre-stained proteins with unknown molecular weights are assigned weights based on the relative mobility of pre-stained standard protein markers.
The kit includes material for six lab groups sharing three gels and requires a vertical electrophoresis apparatus, power supply, micropipets, and polyacrylamide gels, available separately.
Customers Who Bought This Also Bought