ContactUsLinkComponent
Determination of Amino Acid Sequence in a Peptide Lab Activity
Determination of Amino Acid Sequence in a Peptide Lab Activity
# de catalogue 470024-300
Fournisseur:  Kemtec
REFILL KIT DETERMIN OF AMINO ACID SEQ
Determination of Amino Acid Sequence in a Peptide Lab Activity
# de catalogue 470024-300
Fournisseur:  Kemtec
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Determination of Amino Acid Sequence in a Peptide Lab Activity, Refill Kit
  • Cat. no.:
    470024-300
  • Supplier No.:
    8-160R
Spécifications
A propos de cet article
Tie Science History to Science Technique
Follow the path that led biochemist Frederick Sanger to receive two Nobel Prizes with this exciting lab. Everything (except a hairdryer) is included for students to determine amino acid content, composition, and position of terminal groups of the tripeptide arginylglycylphenylalanine from a protein hydrolysate. The activity includes an instructor’s manual, an assessment manual, and student data sheets for four setups to do four different exercises each.
Customers Who Bought This Also Bought