Denoyer-Geppert® Giant Eye Models
Fournisseur: Denoyer-Geppert Science
Certificats
A propos de cet article
The detachable lens magnifies light and shows inverted images, and the removable iris and transparent vitreous body let students discover the delicate structures of the eye.
- The student version is supplied on a dark base, and has fewer identified structures
Students may also dissect the 6× life-size model to study rods, cones and more than other coded structures.
Customers Who Bought This Also Bought