Spécifications
- Length:91 cm
- Width:112 cm
- Description:Basic Biology Charts Set of 20
- Cat. no.:470004-192
- Finish:Unlaminated
- Supplier No.:1996-41
Spécifications
A propos de cet article
These Colorful, Highly Detailed Charts Brilliantly Illuminate Basic Biological, Botanical, And Zoological Concepts.
- Includes tripod and chart head
- Printed on synthetic stock to prevent tears and yellowing
Each chart displays meticulously drawn and labeled illustrations of the internal and external anatomy of the subject. Enlarged sections allow for close-up views of important anatomical features >Life-cycle, reproduction and excretory systems, symbiosis, plant and cell mitosis and meiosis are all covered.
Basic Biology Set of 20 includes: Animal Cell and Plant Cell, Protista, Coelentrates (Cnidaria), Flatworms and Roundworms, Earthworm, Crayfish, Grasshopper, Perch, Frog Anatomy, Animal Mitosis, Animal Meiosis, Roots, Stems, Leaves, Flowers, Monocot and Dicot Seed Germination, Plant Mitosis, Plant Meiosis, Mendelian Heredity, Evolution of The Five Kingdoms.
Complete Biology Set of 35 includes: All of the charts in the Basic set and Honeybee, Clam, Frog Development, Bird Studies, Pathogenic Microorganisms, Green and Brown Algae , Wheat Rust Life Cycle, Liverwort Life Cycle , Moss Life Cycle, Fern Life Cycle , Gymnosperms , Two Kingdom Classification System , Concealing and Warning Adaptations , Symbiosis , Carbon, Oxygen, and Nitrogen Cycles.