Denoyer-Geppert® Cardiopulmonary System
# de catalogue 470004-198
Fournisseur: Denoyer-Geppert Science
Spécifications
- Description:Cardiopulmonary System
- Height:15 cm
- No. of Keyed Items:58
- Cat. no.:470004-198
- Length:42 cm
- Width:28 cm
- Supplier No.:0192-00
- No. of Parts:5
Spécifications
A propos de cet article
This system shows interdependence of circulatory and respiratory systems.
- Wooden Base Mount
- Removable Parts
- Detailed Depictions
- Sectioned Larynx
The life-size heart and lungs are depicted in great detail and vivid color to display all the important structures of both systems involved in pulmonary circulation.
Half of the left lung and the two-part heart are removable to show bifurcation of the trachea, bronchial tree, and pulmonary vessels. The larynx is sectioned, and the right half is removable for closer study. The esophagus and major vessels are also shown.
Customers Who Bought This Also Bought