Denoyer-Geppert® Giant Ear Model
Fournisseur: Denoyer-Geppert Science
Certificats
A propos de cet article
The fluid-filled canals of this model demonstrate the concept of equilibrium.
- Model Enlarged Five Times
- Removable Parts
- Color-Coded Canals
- Models Show How Ears Control Body's Equilibrium
Perfect for anatomy, biology, and speech therapy courses, the 5X-enlarged model presents all ear structures in magnificent detail. Its most unique feature, two semicircular canals filled with fluid, allow students to read the air bubble as if it were a level. The labyrinth and cochlea can be removed to display the vestibule, auditory nerve, and stapes; the cochlea is dissected to show the spiral membrane that aids hearing.
The three canals are color coded. The tympanum, malleus, and the incus are also removable.
Customers Who Bought This Also Bought