Demonstration Bimetallic Strip
# de catalogue 470014-318
Fournisseur: VWR International
Spécifications
- Description:Demonstration Bimetallic Strip
- Size:5 1/4 x 3 1/4 x 2"
- Cat. no.:470014-318
- Supplier No.:PH 0403A
Spécifications
A propos de cet article
Vivid Illustration of Thermoexpansion.
A 12V halogen bulb is mounted below this metallic strip to heat it. Once heated, the strip bends upward and breaks the circuit.
Customers Who Bought This Also Bought