Demonstration Balance Support
Catalog # 470148-748
Supplier: VWR International
Specifications
- Description:Demonstration Balance Support
- Cat. no.:470148-748
- Supplier No.:P10852/3
Specifications
About this item
Add your own lab materials to illustrate torque.
The 'Heart' of our moment of force apparatus, the sturdy notched support, is available either with a knife-edge clamp or alone for you to use the lab materials you already have to complete the apparatus. You will need a meter stick, mass hangers and masses, and several knife-edge clamps, available separately.
Frequently Bought Together