3B Scientific® Deluxe Training Baby
Fournisseur: American 3B Scientific
Certificats
A propos de cet article
Practice Male And Female Procedures On One Model
- Inserts for male or female pediatric training
- 3 year manufacturers
Pediatric nursing trainees can practice basic and critical infant care on this incredibly realistic model with interchangeable male and female sex organs. Fully mobile neck and joints, and internal organs allow students to perform techniques such as tracheotomy care, catheterization, enema insertion, intramuscular and subcutaneous injections, and palpitation of fontanels. Model includes syringe, suction catheter, feeding tube, urinary catheter, urine collection bag, lubricant and carrying bag.
Customers Who Bought This Also Bought