Dehydrated Media for Luria Broth
# de catalogue 470180-630
Fournisseur: IPM SCIENTIFIC, INC.
Numéro CAS:
Spécifications
- Description:Dehydrated Media for Luria Broth, Makes 1 liter
- Cat. no.:470180-630
- Supplier No.:880009
Spécifications
A propos de cet article
Premeasured. Complete nutrient source for growing E. coli cultures.
Sterile, ready-to-use media. Note: Perishable. Please specify delivery date.
Customers Who Bought This Also Bought