ContactUsLinkComponent
Dehydrated Media for Luria Agar
Dehydrated Media for Luria Agar
# de catalogue 470180-628
Fournisseur:  IPM SCIENTIFIC, INC.
Numéro CAS:  
LURIA AGAR DEHYDRATED MEDIA MAKES 1L
Dehydrated Media for Luria Agar
# de catalogue 470180-628
Fournisseur:  IPM SCIENTIFIC, INC.
Numéro CAS:  
Spécifications
  • Description:
    Dehydrated Media for Luria Agar, Makes 1 liter
  • Cat. no.:
    470180-628
  • Supplier No.:
    880008
Spécifications
A propos de cet article
Premeasured. Complete nutrient source for isolating E. coli colonies.
Sterile, ready-to-use media. Note: Perishable. Please specify delivery date.
Customers Who Bought This Also Bought