Decade Inductance Box
Catalog # 470121-490
Supplier: VWR International
Specifications
- Description:Decade Inducatance Box
- Cat. no.:470121-490
- Supplier No.:PH1135F
Specifications
About this item
Investigating Inductance has never been this easy
This sturdy decade box allows a range of inductances from 10 H down to 1 microH. Maximum capacity is 11.11111 H. Housed in a box with seven dials and fitted with 4 mm output plugs, the unit is fully equipped for laboratory use.
Customers Who Bought This Also Bought