Polypropylene, Single Scale Cylinders
# de catalogue 470005-422
Fournisseur: Bel-Art
Spécifications
- Capacity:1000 ml
- Graduation Interval:10 mL
- Description:Polypropylene, Single Scale Cylinders
- Form:Tall
- Cat. no.:470005-422
- Supplier No.:F284580000
Spécifications
A propos de cet article
High Quality, Unbreakable Cylinder.
- Easy to read scale
- Autoclavable up to 135°C
Customers Who Bought This Also Bought