Easy Read Graduated, Single Scale Cylinders
Catalog # 470150-536
Supplier: Avantor
Specifications
- Description:Cylinder, Tenite Plastic, 100 mL x 2 mL, Easy Read Graduate, Single Scale
- Form:Tall
- Cat. no.:470150-536
- Supplier No.:DPS-0013
Specifications
About this item
Accurately calibrated, ideal for physical science work, and a must for special needs students, these cylinders feature large, easy to read graduations and no drip spouts. Made of flexible plastic, they are student proof and offer moderate chemical resistance. The removable base makes for easy storage.
Customers Who Bought This Also Bought