Crucible Holder
# de catalogue 470151-210
Fournisseur: Plasticoid
Spécifications
- Description:Crucible Holder
- Cat. no.:470151-210
- Supplier No.:CWS-M290
Spécifications
A propos de cet article
Made of molded, black rubber with a conical opening 27 mm at the top tapered to 22 mm at the bottom, the crucible holder includes a glass funnel.
- Holder may be used with suction flasks of 250 to 1000 ml capacity
- Holds Gooch and similar crucibles of 35 to 40 mm diameter at the top
Overall length: 60 mm.
Customers Who Bought This Also Bought