Crime Scene 1: The Case of Lois McArthur
# de catalogue 470004-356
Fournisseur: Theta Technologies
Spécifications
- Description:Crime Scene 1: The Case of Lois McArthur
- No. of Lab Groups:12
- Cat. no.:470004-356
- Supplier No.:CS1
Spécifications
A propos de cet article
Students Apply Science Principles and Relate the Conclusion to a Criminal Investigation
- Case-Based Scenario
- Biology and Chemistry Concepts
All the intrigue, mystery, and excitement of an actual case arrives in your lab with this lab activity.
Here’s a chance to relate biology and chemistry to the real life application of criminal investigations.Includes a teacher’s guide, reproducible student instructions, and enough materials for 12 setups.
Tragic suicide or horrible murder? Your students will decide as they perform blood, hair, fiber, fingerprint, and handwriting analysis tests.
Customers Who Bought This Also Bought