Cotton Roll
# de catalogue 470161-446
Fournisseur: MEDLINE INDUSTRIES, INC.
Spécifications
- Description:Cotton Roll, Nonsterile, Absorbent, 1 lb roll
- Cat. no.:470161-446
- Supplier No.:79210006
Spécifications
A propos de cet article
Quantity: 1 lb.
Customers Who Bought This Also Bought