Corundum (Sapphire)
# de catalogue 470026-364
Fournisseur: ADAM MOORE MO
Spécifications
- Description:Corundum (Sapphire) Display Mineral
- Cat. no.:470026-364
- Supplier No.:WARD470026-364
Spécifications
A propos de cet article
Beautiful, blue, hexagonal, sapphire crystals. Average size: 3/8". Madagascar.
Customers Who Bought This Also Bought