Cork Pinning Sheet
# de catalogue 470019-298
Fournisseur: Zandur
Spécifications
- Description:Cork Pinning Sheet
- Cat. no.:470019-298
- Supplier No.:E 324
Spécifications
A propos de cet article
The Preferred Material of Professionals
This high-quality Spanish entomological cork is plain and unpapered. It is supplied in large sheets that can be customized to fit your insect drawers and boxes. Size: 35"L x 23"W x 1/4"H.
Customers Who Bought This Also Bought