Coprolites
Catalog # 470124-458
Supplier: ADAM MOORE MO
Specifications
- Description:Coprolites, Large
- Length:5.08 - 7.62 cm (2 - 3")
- Cat. no.:470124-458
- Supplier No.:535166
Specifications
About this item
- Mammal feces
- Miocene
Customers Who Bought This Also Bought