Ward's® Contour Map Grid
# de catalogue 470029-392
Fournisseur: Plas-Labs
Spécifications
- Description:WARD’S Contour Map Grid
- Cat. no.:470029-392
- Supplier No.:153CGS
Spécifications
A propos de cet article
Strong Enough for Heavy Stream Table Use
Constructed of tough acrylic, the lines of the grid that mark off the 2" squares are permanently etched so they won’t wear off with repeated use. Use the grid with either of WARD’S stream tables to trace contour patterns. Size: 25"L x 24"W x 1/8"H.
Customers Who Bought This Also Bought