Content Area – Graphic Organizers For Science
Catalog # 470007-302
Supplier: Walch Publishing
Specifications
- Description:Graphic organizers for science
- Length:210 mm (8.25")
- Cat. no.:470007-302
- No. of Pages:120
- Supplier No.:05037X
- Width:270 mm (10.6")
Specifications
About this item
Gina Hamilton. You can use this flexible and effective tool with almost any topic in your science curriculum.
- Organizers can be copied for student use and range from sequence chains and concept maps to Venn diagrams, webs, and flow charts
- There are even some organizers that were created specifically for this book
Iincludes a teacher’s guide with tips on how to use each organizer and why graphic organizers help promote learning.
Customers Who Bought This Also Bought