Connecting Wires (BNC to Alligator Clips)
# de catalogue 470122-140
Fournisseur: Avantor
Spécifications
- Description:Connecting wires (BNC to alligator clips)
- Cat. no.:470122-140
- Cord Length:18"
- Supplier No.:4531-C-18
Spécifications
A propos de cet article
Flexible, rubber-covered wire with covered plated steel alligator clips on ends.
Wire is stranded. Set of two leads: one black, one red.
Customers Who Bought This Also Bought