BoneClones® Comparative Maxilla Set
# de catalogue 470030-036
Fournisseur: BONE CLONES, INC.
Small Business Enterprise
Spécifications
- Description:Comparative Maxilla Set
- Cat. no.:470030-036
- Supplier No.:COMP-105
- No. of Parts:3
Spécifications
A propos de cet article
Excellent Comparative Set For Forensic And Anthropological Studies
Discover forensic methods of identifying the race/ethnicity of an individual with this three-piece set. Differences in the maxillary, nasal, dental and palate structures among Asian, African, and European males are clearly illustrated. In the Asian model, the dental arch is rounded and the lingual surface of the incisors is shovel-shaped; the African replica features a hyperbolic dental arch and spatulate incisors; and the European model’s dental arch is parabolic and the incisors are spatulate.
Customers Who Bought This Also Bought