Common Rock Collection
# de catalogue 470104-708
Fournisseur: Avantor
Spécifications
- Description:Common Rock Collection
- No. of Samples:6
- Cat. no.:470104-708
- Supplier No.:WARD470104-708
Spécifications
A propos de cet article
An introductory rock set contains six samples from each of the three major rock types.
- Great Introductory Set
- Hand Cut by Geologists
The numbered samples include: granite, sandstone, limestone, shale, marble, and slate ina compartmented collection box with key list. Size of each: 1" x 1"
Note: Collections are subject to change depending on availability of individual items.
Customers Who Bought This Also Bought