Colorflame Candles
# de catalogue 470145-064
Fournisseur: JOELSON INDUSTRIES
Spécifications
- Description:Colorflame Candles
- Cat. no.:470145-064
- Supplier No.:PEG7632-B
Spécifications
A propos de cet article
Excellent candles for flame tests. Add an element of fun to your chemistry demonstrations with these colorful candles.
- Non-toxic and smokeless
- Biodegradable
The standard birthday candles are anything but traditional. Thes candles burn with red, purple, green, blue, orange, and white flames.
Delivery: Includes 12 candles an 12 clear plastic holders.
Customers Who Bought This Also Bought