ContactUsLinkComponent
Color Filter Paddles
Color Filter Paddles
# de catalogue 470101-612
COLOR FILTER PADDLES 11 FILTERS
Color Filter Paddles
# de catalogue 470101-612
Spécifications
  • Description:
    Color Filter Paddles, 11 Filters
  • Cat. no.:
    470101-612
  • Supplier No.:
    10025
Spécifications
A propos de cet article
Lessons in Optics and Light in the Palm of Your Hand Colorfully illustrate additive and subtractive color mixing, color transmission, absorption and filtering of the different wavelengths of light with this set of primary and secondary color mixing paddles. You can also use the diffraction gratings in this set to view spectra from different light sources. Show students how a spectroscope, colored lights, and even polarized sunglasses work with the contents of this set.
Customers Who Bought This Also Bought