Collision in Two Dimensions
Catalog # 470148-956
Supplier: VWR International
Specifications
- Description:Collision in Two Dimensions
- Cat. no.:470148-956
- Supplier No.:P10505
Specifications
About this item
The Collision in Two Dimensions apparatus is a rich source of data for studying conservation of momentum and conservation of kinetic energy in collisions.
- For Studying a Variety of Scenarios
- Inexpensive
- Eye Screw Works a Plumb Bob
- One Glass Eye Included
- Apparatus Also Compares Elastic and Inelastic Collisions
It is also used to compare elastic and inelastic collisions.
A curved metal track and an adjustable target support from the core of the unit.
Ordering information: This kit includes two steel balls, a glass ball, and an eye screw, which serves as a plumb bob. It requires a C-clamp for mounting on a table, available separately.
Customers Who Bought This Also Bought