Collision Ball Apparatus
Catalog # 470005-398
Supplier: VWR International
Specifications
- Description:Collision Ball Apparatus
- Cat. no.:470005-398
- Supplier No.:COL-12
Specifications
About this item
An easy, inexpensive way to effectively demonstrate conservation of energy and momentum
- Durable
- High Quality
This apparatus includes five steel balls made to collide with each other on a metal track. The metal track is 58 cm long and is supported at both ends.
Customers Who Bought This Also Bought