Spécifications
- Description:Coliscan® EasyGel® Water Monitoring Kit
- No. of Lab Groups:10
- Cat. no.:470107-534
- Supplier No.:CWM10
Spécifications
A propos de cet article
Make Sure the Water is Free of E. coli
- Collect local water samples
- Linked to a real world scenario
- Includes an instruction guide
Coliscan Water Monitoring Kit features Easygel, an agar replacement. Each kit has materials for 10 tests. The kit includes Coliscan Easygel bottles, Easygel petri dishes, 30mL collection bottles, 1mL Droppers, instruction guide, and Coliscan Interpretation Guide.