Eisco® Cockroach Model
# de catalogue 470111-056
Fournisseur: VWR International
Spécifications
- Description:Cockroach
- Height:8 cm
- Cat. no.:470111-056
- Length:46 cm
- Width:20 cm
- Supplier No.:ZM0044
- No. of Parts:1
Spécifications
A propos de cet article
We can learn a lot about insects from this common pest. A enlarged dissection of a cockroach shows all internal organs, including the salivary glands.
Mounted on a plastic base, the model includes a key card for all numbered structures.
Customers Who Bought This Also Bought