Clear Polystyrene Spot Plates
# de catalogue 470005-972
Fournisseur: PRECISION MOULDED POLYMERS LTD
Spécifications
- Description:Clear Polystyrene Spot Plates
- Cat. no.:470005-972
- Supplier No.:A-M 9000
Spécifications
A propos de cet article
Offering more depressions than other spot plates, these plates are suitable for most lab applications. Made of clear polystyrene with an attached measuring scoop.
- 15 depressions measure 2 ml deep
- Each plate measures 140×80 mm with 15 depressions
Packaging: Package of 10.
Caution: Not recommended for use with strong solvents.
Customers Who Bought This Also Bought