ContactUsLinkComponent
Classroom DNA Electrophoresis LabStation
Classroom DNA Electrophoresis LabStation
# de catalogue 470122-878
Fournisseur:  Edvotek
DNA ELECTROPHORESIS
Classroom DNA Electrophoresis LabStation
# de catalogue 470122-878
Fournisseur:  Edvotek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Classroom DNA Electrophoresis LabStation™
  • Cat. no.:
    470122-878
  • Supplier No.:
    5062
Spécifications
A propos de cet article
High Quality Supplies for Your Classroom
  • Quality materials for a small class
  • Fixed volume micropipettes
  • Fingerprinting kit
With enough materials for your whole class to participate in DNA electrophoresis experiments, this LabStation contains high quality equipment to support up to 24 students.
Kit includes: 1 Electrophoresis Apparatus, 1 Power Source, 2 Fixed Volume MiniPipets (40 μL), 96 Micropipette Tips, DNA Fingerprinting Classroom Kit.
Customers Who Bought This Also Bought