Bull's Eye Level
# de catalogue 470122-634
Fournisseur: Geier & Bluhm
Spécifications
- Description:Bull's Eye Level
- Cat. no.:470122-634
- Supplier No.:2-12000
Spécifications
A propos de cet article
Perfect for leveling ripple tanks, before filling with water, this circular spirit level is enclosed in a chrome plated brass case.
Ø: 3 cm.
Customers Who Bought This Also Bought