Preserved Ciona
Fournisseur: Avantor
Certificats
A propos de cet article
Formalin preserved ciona. Available in jars.
- Tunicate anatomy
- Developemental biology
- Genomics
Ciona (also called sea vase) allow for the study of tunicates. This invertebrate is great for studying developmental biology.
Customers Who Bought This Also Bought