Chromosomes and Genes in Action Microslide
Fournisseur: SUPERTEK SCIENTIFIC LLC
Certificats
A propos de cet article
Investigate Genetic Material
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes DNA molecule (em, 55,000X), Coiling in chromosomes (wm, 2,000X), Lampbrush chromosomes (pc, 700X), Loop of lampbrush chromosomes (em, 9,000X/15,000X),
Giant chromosomes (wm, 1,000X), Chromosomes puffs, stained (250X), Chromosome puffs, tagged (250X), Nuclear pores (em, 50,000X), and Ribosomes (em, 85,000X).
Box of 15 Sets also includes Background subject information and Reproducible student worksheets with answer keys.
Giant chromosomes (wm, 1,000X), Chromosomes puffs, stained (250X), Chromosome puffs, tagged (250X), Nuclear pores (em, 50,000X), and Ribosomes (em, 85,000X).
Box of 15 Sets also includes Background subject information and Reproducible student worksheets with answer keys.
Customers Who Bought This Also Bought