Chocolate Agar, Tubes
# de catalogue 470180-644
Fournisseur: Culture Media & Supplies
Numéro CAS:
Spécifications
- Description:Prepared Chocolate Agar, Tubes
- Cat. no.:470180-644
- Supplier No.:WARD470180-644
Spécifications
A propos de cet article
Supplemented with X and V factors
For cultivation of Neisseria and Haemophilus species. Package of 12 tubes supplied.
Customers Who Bought This Also Bought