Chlorellopsis coloniata (Eocene)
# de catalogue 470027-510
Fournisseur: Avantor
Spécifications
- Description:Chlorellopsis coloniata
- Cat. no.:470027-510
- Supplier No.:WARD470027-510
Spécifications
A propos de cet article
Freshwater algal structures in chert. Average size: 3" x 4".
Customers Who Bought This Also Bought