Chemistry Hardware Assortment Set
# de catalogue 470126-938
Fournisseur: United Scientific Supplies
Spécifications
- Description:Chemistry Hardware Assortment Set, 1 Lab Station
- No. of Lab Groups:1; 2-4 Users per Set
- Cat. no.:470126-938
- Supplier No.:HRDKIT2-C
Spécifications
A propos de cet article
The Basic Tools for an Efficient Chemistry Lab.
- Includes all the tools needed to perform hundreds of chemistry experiments and procedures in a general chemistry lab
Includes (1) of each of the following: Beaker tong, Burette clamp, Burner tripod, Pinchcock clamp, Right angle clamp holder, Clay pipe triangle, Crucible tong, 4" ring support with clamp, Ring stand (5" x 8" base), Six-tube test tube rack, Test tube clamp, Wire gauze, 5" x 5".
Customers Who Bought This Also Bought