Chemical Spill Tray
Fournisseur: SCIENTIFIC PLASTICS
Certificats
A propos de cet article
Safely Contain Spills!
- Available in three sizes
- All trays are 2" deep
One-piece, rotationally-molded polyolefin trays protect cabinets and shelving from acid spills. Corners are rounded for easy cleaning, and walls are 4.8mm (3/16") thick for rigidity. Trays are dishwasher safe and autoclavable.
Customers Who Bought This Also Bought