Charger for Killing Jar
# de catalogue 470018-456
Fournisseur: Ward's Science
Spécifications
- Description:Charger for Killing Jar
- Cat. no.:470018-456
- Supplier No.:EE0068-KIT
Spécifications
A propos de cet article
Includes Squeeze Bottle for Convenient Application
Ethyl acetate’s relatively low toxicity to humans, yet quick action on insects makes this a recommended killing agent. The package includes a refillable 1 oz. plastic squeeze bottle with 100 mL of ethyl acetate. With MSDS. Note: Shipping and drug restrictions apply. Sold on institutional purchase order only.
Customers Who Bought This Also Bought