Cerebellum and Choroid Plexus Slide
# de catalogue 470182-722
Fournisseur: Avantor
Spécifications
- Description:Cerebellum & Choroid Plexus
- Cat. no.:470182-722
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-722
- Orientation:Section
Spécifications
A propos de cet article
Region of the Brain
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Section through portion of cerebellum and medulla oblongata in the region of the fourth ventricle.
Customers Who Bought This Also Bought