Cerebellum, Mammal Slide
# de catalogue 470177-606
Fournisseur: Avantor
Spécifications
- Description:Cerebellum, Mammal
- Cat. no.:470177-606
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470177-606
- Orientation:Section
Spécifications
A propos de cet article
Illustrates Structures of the Brain
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Demonstrates molecular and granular layers of the cortex and Purkinje cells.
Customers Who Bought This Also Bought