Ceramic Ring Magnets
Catalog # 470105-290
Supplier: Master Magnetics
Specifications
- Description:Ceramic Ring Magnets
- Cat. no.:470105-290
- Supplier No.:CR162
Specifications
About this item
These ceramic ring magnets have an inside diameter of 35 mm and an outside diameter of 45 mm.
They are approximately 6.4 mm thick and are magnetized parallel with the thickness.
Packaging: Package of 16.
Customers Who Bought This Also Bought